Assay Details
| Gene Symbol: Krt17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511335.2 | XP_003511383.1 | 7 - 8 | 1205 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt17| Gene Symbol: Krt17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511335.2 | XP_003511383.1 | 7 - 8 | 1205 | 84 | |
Set Membership: |
|