Assay Details
| Gene Symbol: LOC100754866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612479.1 | XP_007610669.1 | 7 - 8 | 877 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100754866| Gene Symbol: LOC100754866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007612479.1 | XP_007610669.1 | 7 - 8 | 877 | 108 | |
Set Membership: |
|