Assay Details
| Gene Symbol: S100a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647669.1 | XP_007645859.1 | 2 - 3 | 445 | 90 | |
| XM_007647670.1 | XP_007645860.1 | 2 - 3 | 223 | 90 | ||
| XM_007647671.1 | XP_007645861.1 | 2 - 3 | 170 | 90 | ||
| XM_007647672.1 | XP_007645862.1 | 1 - 2 | 159 | 90 | ||
| XM_007647673.1 | XP_007645863.1 | 2 - 3 | 253 | 90 | ||
| GenBank mRNA | JI881046.1 | - | 1 - 2 | 229 | 90 | |
| JP061800.1 | - | 1 - 2 | 229 | 90 | ||
More Information
Set Membership: |
|

S100a10
Genomic Map