Assay Details
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505010.2 | XP_003505058.1 | 3 - 4 | 266 | 63 | |
| XM_007645461.1 | XP_007643651.1 | 3 - 4 | 266 | 63 | ||
| XM_007645463.1 | XP_007643653.1 | 3 - 4 | 266 | 63 | ||
| XM_007645464.1 | XP_007643654.1 | 3 - 4 | 266 | 63 | ||
More Information
Set Membership: |
|

Cd72
Genomic Map