Assay Details
Gene Symbol: Calhm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512142.2 | XP_003512190.2 | 3 - 4 | 574 | 60 | |
XM_007652627.1 | XP_007650817.1 | 3 - 4 | 906 | 60 | ||
XM_007652628.1 | XP_007650818.1 | 3 - 4 | 676 | 60 | ||
XM_007652629.1 | XP_007650819.1 | 3 - 4 | 1015 | 60 | ||
XM_007652630.1 | XP_007650820.1 | 3 - 4 | 1765 | 60 | ||
XM_007652631.1 | XP_007650821.1 | 2 - 3 | 1567 | 60 | ||
XM_007652632.1 | XP_007650822.1 | 2 - 3 | 1337 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
