Assay Details
| Gene Symbol: Bpgm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513040.2 | XP_003513088.1 | 2 - 3 | 789 | 73 | |
| GenBank mRNA | JI876806.1 | - | 1 - 2 | 628 | 73 | |
More Information
Set Membership: |
|

Bpgm
Genomic Map