Assay Details
Gene Symbol: Slc9a3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643172.1 | XP_007641362.1 | 2 - 3 | 770 | 57 | |
GenBank mRNA | AF307992.1 | - | 1 - 2 | 51 | 57 | |
JI887063.1 | - | 2 - 3 | 293 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
