Assay Details
Gene Symbol: LOC100759042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642550.1 | XP_007640740.1 | 2 - 3 | 565 | 70 | |
XM_007642551.1 | XP_007640741.1 | 3 - 4 | 627 | 70 | ||
XM_007642552.1 | XP_007640742.1 | 3 - 4 | 533 | 70 | ||
XM_007642553.1 | XP_007640743.1 | 2 - 3 | 206 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
