Assay Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495042.2 | XP_003495090.1 | 6 - 7 | 982 | 69 | |
| GenBank mRNA | JI862079.1 | - | 6 - 7 | 964 | 69 | |
| JP054034.1 | - | 6 - 7 | 952 | 69 | ||
More Information
Set Membership: |
|

Ccnb2
Genomic Map