Assay Details
| Gene Symbol: Ahsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499661.2 | XP_003499709.1 | 7 - 8 | 947 | 63 | |
| GenBank mRNA | JI892176.1 | - | 7 - 8 | 945 | 63 | |
| JP038454.1 | - | 7 - 8 | 947 | 63 | ||
More Information
Set Membership: |
|

Ahsa1
Genomic Map