Assay Details
Gene Symbol: Isx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515099.1 | XP_003515147.1 | 3 - 4 | 492 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
