Assay Details
| Gene Symbol: Mapre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502308.2 | XP_003502356.1 | 4 - 5 | 520 | 84 | |
| XM_007642911.1 | XP_007641101.1 | 4 - 5 | 518 | 84 | ||
| XM_007642912.1 | XP_007641102.1 | 4 - 5 | 497 | 84 | ||
| GenBank mRNA | JI882651.1 | - | 3 - 4 | 482 | 84 | |
| JP055508.1 | - | 3 - 4 | 581 | 84 | ||
More Information
Set Membership: |
|

Mapre1
Genomic Map