Assay Details
| Gene Symbol: LOC103162924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649698.1 | XP_007647888.1 | 2 - 3 | 595 | 67 | |
| XM_007649699.1 | XP_007647889.1 | 2 - 3 | 595 | 67 | ||
More Information
Set Membership: |
|

LOC103162924
Genomic Map