Assay Details
| Gene Symbol: LOC103162924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649698.1 | XP_007647888.1 | 3 - 4 | 702 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103162924| Gene Symbol: LOC103162924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649698.1 | XP_007647888.1 | 3 - 4 | 702 | 75 | |