Assay Details
| Gene Symbol: LOC103162924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649700.1 | XP_007647890.1 | 1 - 2 | 457 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103162924| Gene Symbol: LOC103162924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649700.1 | XP_007647890.1 | 1 - 2 | 457 | 79 | |