Assay Details
Gene Symbol: Synpo2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496719.1 | XP_003496767.1 | 3 - 4 | 771 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
