Assay Details
| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646909.1 | XP_007645099.1 | 1 - 2 | 243 | 55 | |
| GenBank mRNA | JP065185.1 | - | 243 | 55 | ||
More Information
Set Membership: |
|

Sec11c
Genomic Map