Assay Details
Gene Symbol: LOC100771856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498154.2 | XP_003498202.1 | 6 - 7 | 807 | 95 | |
XM_003498155.2 | XP_003498203.1 | 6 - 7 | 816 | 95 | ||
XM_007622986.1 | XP_007621176.1 | 6 - 7 | 828 | 95 | ||
XM_007622992.1 | XP_007621182.1 | 6 - 7 | 825 | 95 | ||
XM_007622997.1 | XP_007621187.1 | 7 - 8 | 836 | 95 | ||
XM_007623015.1 | XP_007621205.1 | 6 - 7 | 813 | 95 | ||
XM_007623030.1 | XP_007621220.1 | 6 - 7 | 804 | 95 | ||
GenBank mRNA | JI882472.1 | - | 2 - 3 | 198 | 95 | |
JP039114.1 | - | 2 - 3 | 192 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
