Assay Details
| Gene Symbol: Hnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510615.2 | XP_003510663.1 | 3 - 4 | 809 | 61 | |
| XM_007651237.1 | XP_007649427.1 | 3 - 4 | 809 | 61 | ||
| XM_007651238.1 | XP_007649428.1 | 3 - 4 | 731 | 61 | ||
| XM_007651239.1 | XP_007649429.1 | 3 - 4 | 731 | 61 | ||
More Information
Set Membership: |
|

Hnf1b
Genomic Map