Assay Details
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643240.1 | XP_007641430.1 | 3 - 4 | 381 | 59 | |
GenBank mRNA | AF320819.1 | - | 2 - 3 | 249 | 59 | |
JI869101.1 | - | 2 - 3 | 161 | 59 | ||
JP037417.1 | - | 2 - 3 | 161 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
