Assay Details
| Gene Symbol: Pla2g1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645289.1 | XP_007643479.1 | 1 - 2 | 157 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g1b| Gene Symbol: Pla2g1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645289.1 | XP_007643479.1 | 1 - 2 | 157 | 70 | |
Set Membership: |
|