Assay Details
| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651232.1 | XP_007649422.1 | 3 - 4 | 377 | 55 | |
| GenBank mRNA | JI862907.1 | - | 3 - 4 | 377 | 55 | |
More Information
Set Membership: |
|

Eif3f
Genomic Map