Assay Details
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654984.1 | XP_007653174.1 | 1 - 2 | 119 | 69 | |
GenBank mRNA | JP039567.1 | - | 241 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
