Assay Details
Gene Symbol: Sohlh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502952.1 | XP_003503000.1 | 9 - 10 | 998 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
