Assay Details
| Gene Symbol: Prmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514079.2 | XP_003514127.1 | 8 - 9 | 926 | 60 | |
| XM_003514080.2 | XP_003514128.1 | 9 - 10 | 964 | 60 | ||
| GenBank mRNA | JI894499.1 | - | 7 - 8 | 827 | 60 | |
| JP065048.1 | - | 9 - 10 | 959 | 60 | ||
More Information
Set Membership: |
|

Prmt1
Genomic Map