Assay Details
| Gene Symbol: Senp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007608341.1 | XP_007606531.1 | 1 - 2 | 734 | 65 | |
| GenBank mRNA | JI891235.1 | - | 1 - 2 | 636 | 65 | |
| JP051630.1 | - | 1 - 2 | 46 | 65 | ||
More Information
Set Membership: |
|

Senp3
Genomic Map