Assay Details
Gene Symbol: Ovca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647841.1 | XP_007646031.1 | 1 - 2 | 251 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
