Assay Details
| Gene Symbol: LOC100769854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641744.1 | XP_007639934.1 | 2 - 3 | 911 | 79 | |
| GenBank mRNA | JP065714.1 | - | 2 - 3 | 198 | 79 | |
More Information
Set Membership: |
|

LOC100769854
Genomic Map