Assay Details
| Gene Symbol: Rps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508652.2 | XP_003508700.1 | 3 - 4 | 414 | 114 | |
| GenBank mRNA | JI873772.1 | - | 3 - 4 | 289 | 114 | |
| JP040262.1 | - | 3 - 4 | 289 | 114 | ||
More Information
Set Membership: |
|

Rps11
Genomic Map