Assay Details
Gene Symbol: LOC100760025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507568.2 | XP_003507616.1 | 2 - 3 | 122 | 98 | |
GenBank mRNA | JP062469.1 | - | 1 | 115 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
