Assay Details
Gene Symbol: Tspyl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644587.1 | XP_007642777.1 | 2 - 3 | 569 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
