Assay Details
| Gene Symbol: Lsm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502589.2 | XP_003502637.1 | 3 - 4 | 188 | 94 | |
| XM_007643230.1 | XP_007641420.1 | 3 - 4 | 199 | 94 | ||
| GenBank mRNA | JI886093.1 | - | 3 - 4 | 198 | 94 | |
| JP055721.1 | - | 3 - 4 | 198 | 94 | ||
More Information
Set Membership: |
|

Lsm7
Genomic Map