Assay Details
Gene Symbol: LOC100767934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498736.2 | XP_003498784.2 | 1 | 217 | 92 | |
GenBank mRNA | JI881212.1 | - | 2 - 3 | 217 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
