Assay Details
Gene Symbol: Nutf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504288.2 | XP_003504336.1 | 3 - 4 | 218 | 101 | |
XM_007644855.1 | XP_007643045.1 | 3 - 4 | 279 | 101 | ||
GenBank mRNA | JI886388.1 | - | 3 - 4 | 490 | 101 | |
JP052680.1 | - | 3 - 4 | 490 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
