Assay Details
| Gene Symbol: Hnrnpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007626195.1 | XP_007624385.1 | 1 | 234 | 98 | |
| GenBank mRNA | JP063658.1 | - | 1 | 234 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnrnpa3| Gene Symbol: Hnrnpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007626195.1 | XP_007624385.1 | 1 | 234 | 98 | |
| GenBank mRNA | JP063658.1 | - | 1 | 234 | 98 | |
Set Membership: |
|