Assay Details
Gene Symbol: LOC100751951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507230.1 | XP_003507278.1 | 1 - 2 | 246 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
