Assay Details
Gene Symbol: LOC100766952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497493.2 | XP_003497541.1 | 36 - 37 | 5587 | 110 | |
XM_007613034.1 | XP_007611224.1 | 36 - 37 | 5575 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
