Assay Details
| Gene Symbol: Amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645406.1 | XP_007643596.1 | 1 | 753 | 93 | |
| GenBank mRNA | JP045107.1 | - | 1 | 749 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amd1| Gene Symbol: Amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645406.1 | XP_007643596.1 | 1 | 753 | 93 | |
| GenBank mRNA | JP045107.1 | - | 1 | 749 | 93 | |
Set Membership: |
|