Assay Details
| Gene Symbol: Arpp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495016.2 | XP_003495064.1 | 1 - 2 | 142 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arpp19| Gene Symbol: Arpp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495016.2 | XP_003495064.1 | 1 - 2 | 142 | 119 | |
Set Membership: |
|