Assay Details
| Gene Symbol: Ube2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498489.2 | XP_003498537.2 | 3 - 4 | 292 | 104 | |
| GenBank mRNA | JI880631.1 | - | 2 - 3 | 118 | 104 | |
| JP045035.1 | - | 3 - 4 | 292 | 104 | ||
More Information
Set Membership: |
|

Ube2b
Genomic Map