Assay Details
Gene Symbol: Lrif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652869.1 | XP_007651059.1 | 4 | 2423 | 87 | |
XM_007652871.1 | XP_007651061.1 | 4 | 2328 | 87 | ||
XM_007652872.1 | XP_007651062.1 | 4 | 2415 | 87 | ||
XM_007652873.1 | XP_007651063.1 | 4 | 1040 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
