Assay Details
| Gene Symbol: Ywhaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504324.2 | XP_003504372.1 | 1 | 122 | 108 | |
| GenBank mRNA | JI867018.1 | - | 1 | 266 | 108 | |
| JP048141.1 | - | 1 | 266 | 108 | ||
More Information
Set Membership: |
|

Ywhaz
Genomic Map