Assay Details
| Gene Symbol: Tsku | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507080.2 | XP_003507128.1 | 2 | 819 | 80 | |
| XM_007647637.1 | XP_007645827.1 | 3 | 768 | 80 | ||
| XM_007647638.1 | XP_007645828.1 | 3 | 876 | 80 | ||
| XM_007647639.1 | XP_007645829.1 | 2 | 695 | 80 | ||
| XM_007647641.1 | XP_007645831.1 | 2 | 617 | 80 | ||
More Information
Set Membership: |
|

Tsku
Genomic Map