Assay Details
| Gene Symbol: Chst12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496297.2 | XP_003496345.1 | 2 | 1521 | 73 | |
| XM_007648934.1 | XP_007647124.1 | 2 | 1517 | 73 | ||
More Information
Set Membership: |
|

Chst12
Genomic Map