Assay Details
Gene Symbol: Sertad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498104.2 | XP_003498152.1 | 2 | 419 | 82 | |
XM_007622671.1 | XP_007620861.1 | 2 | 444 | 82 | ||
XM_007622677.1 | XP_007620867.1 | 2 | 497 | 82 | ||
XM_007622678.1 | XP_007620868.1 | 2 | 423 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
