Assay Details
Gene Symbol: Lsm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500864.2 | XP_003500912.1 | 2 | 621 | 125 | |
XM_007641620.1 | XP_007639810.1 | 2 | 530 | 125 | ||
XM_007641621.1 | XP_007639811.1 | 2 | 524 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
