Assay Details
| Gene Symbol: Ube2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502820.2 | XP_003502868.1 | 4 | 622 | 109 | |
| XM_007643458.1 | XP_007641648.1 | 4 | 620 | 109 | ||
| XM_007643459.1 | XP_007641649.1 | 3 | 517 | 109 | ||
| GenBank mRNA | JI879197.1 | - | 4 | 621 | 109 | |
| JP052441.1 | - | 4 | 622 | 109 | ||
More Information
Set Membership: |
|

Ube2l3
Genomic Map