Assay Details
Gene Symbol: Tlr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246762.1 | NP_001233691.1 | 3 | 2706 | 125 | |
XM_007649674.1 | XP_007647864.1 | 2 | 2222 | 125 | ||
GenBank mRNA | AF153676.1 | - | 3 | 2706 | 125 | |
JI881679.1 | - | 3 | 2762 | 125 | ||
JP060647.1 | - | 3 | 2762 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
