Assay Details
Gene Symbol: Rax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507021.1 | XP_003507069.1 | 2 | 221 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
