Assay Details
Gene Symbol: Itpripl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515617.2 | XP_003515665.1 | 1 | 1278 | 128 | |
XM_007607754.1 | XP_007605944.1 | 1 | 1278 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
